Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fluorescent Compatible Sample Buffer (4X, non-reducing)

Catalog No. LC2570
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2570 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2570 Supplier Invitrogen™ Supplier No. LC2570
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen Fluorescent Compatible Sample Buffer is for use in SDS-polyacrylamide gel electrophoresis (SDS-PAGE).

It is optimized for use in fluorescent western blotting as it does not interfere in fluorescent channels, unlike other standard sample buffers. Fluorescent Compatible Sample Buffer is non-reducing but may also be used under denaturing conditions.

• Non-reducing—ready-to-use for non-reducing SDS-PAGE, or the preferred type and amount of reducing agent (e.g., DTT) can be added to produce reducing conditions
• Lane marker dyes—visible markers of the dye front
• Convenient—stable at room temperature, enabling storage at the bench where electrophoresis is performed
• Concentrated—4X formulation provides more versatility than traditional 2X buffers
• Ideal for fluorescent detection—formulated to prevent auto fluorescence during fluorescent detection

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Buffer Sample Loading Buffers
Concentration 4X
Detection Method Fluorescence
Gel Type Tricine, Tris-Glycine, SDS-PAGE, Bis-Tris, Bis-Tris Plus, Tris-Acetate
Gel Compatibility SDS-PAGE Gels
Quantity 5 mL
Shipping Condition Room Temperature
Product Type Fluorescent Compatible Sample Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.