Learn More
Abcam Fluorescent Exosome Standard, B16F10 Mouse Cell Line, Liquid, 100 μL
Description
- Source: B16F10 Mouse cell line (Mouse melanoma cell line)
- Form: Liquid
- Applications: Fluorescence Microscopy, EM, Flow Cytometry
- Storage: Store at -20°C
- Shipping: Blue Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Exosomes (Mouse) |
| Shipping Condition | Blue Ice |
| Form | Liquid |
| Reactivity | Mouse |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Cross Reactivity | Mouse |
| Quantity | 100 μL |
| For Use With (Application) | Fluorescence microscopy, EM, Flow cytometry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.