Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Fluorescent Exosome Standards (Human Serum)
SDP

Catalog No. NB397919 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397919 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397919 Supplier R&D Systems™ Supplier No. NBP311693
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

Specifications

Content And Storage Store at -20°C in the dark. Avoid freeze-thaw cycles.
For Use With (Application) Flow Cytometry, Electron Microscopy
Excitation/Emission The excitation maximum of fluorescent exosome standards is 500 nm - 650 nm and emission maximum is 510 - 665 nm.
Format Human biofluids of healthy donors
Product Line Novus
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.