Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fluorescent Human ES/iPS Cell Characterization Kit

Catalog No. SCR078 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR078 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR078 Supplier MilliporeSigma™ Supplier No. SCR078
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Fast Red Violet solution: One 15mL bottle
  • Napthol AS-BI phosphate solution. One 15mL bottle
  • Mouse anti-Oct-4 (POU5f1), clone 7F9.2, Alexa Fluor™ 488 conjugate. One vial containing 50μL of 0.5mg/mL conjugated monoclonal antibody
  • Mouse anti-Sox-2, clone 10H9.1, Cy3 conjugate. One vial containing 50μL of 0.5mg/mL conjugated monoclonal antibody
  • Mouse anti-Nanog, clone 7F7.1, Alexa Fluor™ 488 conjugate. One vial containing 50μL of 0.5mg/mL conjugated monoclonal antibody
  • Mouse anti-TRA-1-60, clone TRA-1-60, Cy3 conjugate. One vial containing 50μL of 0.5mg/mL conjugated monoclonal antibody
  • Mouse anti-TRA-1-81, clone TRA-1-81, Cy3 conjugate. One vial containing 50μL of 0.5mg/mL conjugated monoclonal antibody
  • DAPI, 100μL. One vial containing 100μL volume

Contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of human Embryonic stem & induced pluripotent Stem cells

Contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of human Embryonic stem and induced pluripotent Stem cells
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2° to 8°C until expiration date of kit
For Use With (Application) Immunocytochemistry
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.