Learn More
STEMCELL Technologies Mitochondrial Tracking Dye, Blue, Size: 500 Tests
SDP
Supplier:  STEMCELL Technologies 1000996
Encompass_Preferred
Rapidly label mitochondria in proliferating and non-proliferating cells with Mitochondrial Tracking Dye, Blue. Using a quick and simple staining protocol, this dye requires minimal hands-on time while promoting photostability and long retention times. Mitochondrial Tracking Dye, Blue is hydrophobic, allowing it to easily cross a live cell membrane and accumulate in mitochondria. Upon depolarization, the dye relocalizes to the cytoplasm and becomes non-specific, but still retains its blue fluorescence with an emission maximum of 469 nm. Mitochondrial Tracking Dye, Blue is well suited for high-throughput screening and is compatible with fluorescence microscopy. It is also suitable for multi-labeling experiments such as microplate assays and flow cytometry. Use mitochondrial tracking dyes to quantify mitochondrial activity, localization, and abundance. For more information about this product, visit www.stemcell.com.
Catalog No. 50-225-0351
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.