Learn More
Fluorescent Mineral Collection, Standard
SDPSCI_ED
Catalog No. S45074
Click to view available options
Quantity:
15 Specimens
Catalog No. Quantity
S45074 15 Specimens
1 options
Catalog No. S45074 Supplier Geoscience Industries Supplier No. 1571000
Suitable for geology and general science courses!
For demonstrating the response of different minerals to both long-wave and short-wave ultraviolet radiation. Each is numbered individually to match an identification key. Housed in a compartmented tray.
Order Info
Contains 15 specimens, each measuring approximately 1 x 1 in. .
Specifications
Quantity 15 Specimens
Specimen Size 1 x 1 in.