Learn More
Description
Biochem/physiol Actions
The metabolism and excretion of the xenobiotic compound 2-fluoroaniline is important due to human exposure in manufacturing. It is found to be very efficiently metabolized, primarily by 4-hydroxylati
Specifications
Specifications
| Chemical Name or Material | 2-Fluoroaniline |
| Packaging | Glass Bottle |
| CAS | 348-54-9 |
| Physical Form | Liquid |
| Linear Formula | FC6H4NH2 |
| MDL Number | MFCD00007642 |
| Beilstein | 1524219 |
| Synonym | 1-Amino-2-fluorobenzene |
| Molecular Weight (g/mol) | 111.12 |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.