Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 5 ppm F- (Premixed with TISAB/Color Coded Blue), Ricca Chemical
SDP

Catalog No. 316432 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3164-32 1 L
3164-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3164-32 Supplier Ricca Chemical Company Supplier No. 316432
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Fluoride Standard
Packaging Natural Poly Bottle
CAS 7681-49-4, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7, 7732-18-5
Concentration or Composition Notes 5ppm F-
CAS Min % 93.0, 1.92, 1.61, 1.56, 0.11, 4.0E-4
CAS Max % 97.0, 2.0, 1.68, 1.63, 0.11, 4.0E-4
Equivalence 1 mL = 0.005 mg F-
Physical Form Liquid
Color Blue
Melting Point Approximately 0°C
Quantity 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.