Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluoro-Max™ Fluorescent Carboxylate-Modified Particles

Catalog No. 09981106
Change view
Click to view available options
Diameter (Metric) Mean:
0.1 μm
0.1, 0.2, 0.3 μm
0.2 μm
0.3 μm
Quantity:
1 mL
5 mL
3 x 1 mL
100 mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Mean Quantity
09-981-106 0.2 μm 5 mL
09-981-103 0.1 μm 5 mL
09-981-104 0.1 μm 100 mL
09-981-102 0.1 μm 1 mL
09-981-107 0.2 μm 100 mL
09-981-105 0.2 μm 1 mL
09-981-109 0.3 μm 5 mL
09-981-110 0.3 μm 100 mL
09-981-108 0.3 μm 1 mL
09-981-900 0.1, 0.2, 0.3 μm 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 09-981-106 Supplier Thermo Scientific™ Supplier No. 93470520010150
Add to Cart
Add to Cart

Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence.

Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence with Thermo Scientific™ Fluoro-Max Fluorescent Carboxylate-Modified Particles. Made by dyeing Thermo Scientific™ OptiLink™ carboxylate-modified particles with europium chelate, particles have been specifically designed for membrane or automated fluorometric-based applications. With an extremely broad Stokes shift, Fluoro-Max europium chelate particles help prevent nonspecific fluorescence interference.

Fluoro-Max Fluorescent Particles are made from carboxylate-modified polystyrene particles containing encapsulated europium chelate dye. They are available in standard 0.1, 0.2, or 0.3μm diameters. When these particles excite at 333nm and emit at 613nm, they do so with a very long lifetime of approximately 0.5 milliseconds. In fact, this is about 10,000 to 100,000 times the lifetime of most fluorophores. The extended lifetime of europium chelate particles allows them to be used in a wide range of time resolved fluorescence applications. In addition, they can be counted on to lower the detection limits of fluorescent assays severalfold as they eliminate background interference from relatively short-lived matrix fluorescence.
  • Internally dyed to ensure maximum color brilliance and color saturation, resulting in greater sensitivity and enhanced readability of the assay. Eliminates problems associated with enzyme-based assays
  • Encapsulated dye prevents leaching in aqueous media, and provides optimum surface immunoreactivity
  • Dyeing process causes little or no inhibition of adsorption or covalent couping of immunoglobulin G, resulting in superior binding of protein to the particles
  • Suitable for a variety of applications such as clinical diagnostics, immuno/histological studies and research

Recommended for:

Clinical diagnostics, molecular biology and lateral flow, turbidimetric, and slide aggulation assays

TRUSTED_SUSTAINABILITY

Specifications

Additive 0.05% Sodium Azide
Binding Property Carboxylate-modified
Concentration 1% solids
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Shipment at ambient temperature. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Contents Fluorescent red dyed polystyrene microspheres in water
Color White
Material Polystyrene or Polystyrene with Coploymer Grafted Surface
Product Type Fluroescent Carboxylate-Modified Particle
Dye Type Europium Chelate
Includes Package Insert Sheet containing Certicate of Analysis
Parking Area 20 A²/group
Particle Density 1.06g/cm³
Quantity 5 mL
Stability Product stable for a minimum of 36 months
Surface Modification Carboxylate-Modified
Uniformity <5% CV
Diameter (Metric) Mean 0.2 μm
Diameter (Metric) Nominal 0.2 μm
Diameter (Metric) 0.2 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.