Learn More
Thermo Scientific™ Fluoro-Max Green and Red Dry Fluorescent Particles
Description
Improve sensitivity and detectability for analytical methods with Thermo Scientific™ Fluoro-Max Green and Red Dry Fluorescent Particles, polymer particles that are internally dyed and feature bright, distinct colors.
Note:
Spectral information is approximate and for reference only. The spectral properties of the dyes are dependent on their concentration and physical environment. The exact excitation and emission maxima may vary depending on the size and composition of the particles.
Recommended for:
- Filter testing
- Flow tracing
- Biological research
Specifications
Specifications
| Additive | May contain trace amount of dispersant |
| Content And Storage | Store at room temperature in a dry area. Store upright and keep bottle tightly sealed. Respiratory protection from dust is recommended. |
| Contents | Dry dyed polystyrene (DVB) microspheres |
| Color | Green |
| Material | Polystyrene Divinylbenzene (PS-DVB) |
| Product Type | Fluorescent Particle |
| Dye Type | Firefli Fluorescent Green (468/508nm) |
| Includes | Package Insert Sheet |
| No. of Cells | 2.83 x 107/gram |
| Particle Density | 1.05g/cm³ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.