Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Fluorocinnamic Acid 98%, Thermo Scientific™

Catalog No. AC172680010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172680010 1 g
AC172680050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172680010 Supplier Thermo Scientific Chemicals Supplier No. 172680010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-90-3
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
InChI Key QONCEXMULRJPPY-VURMDHGXSA-N
Synonym alpha-fluorocinnamic acid, .alpha.-fluorocinnamic acid, cinnamic acid, alpha-fluoro, z-alpha-fluorocinnamic acid, 2-propenoic acid, 2-fluoro-3-phenyl, z-2-fluoro-3-phenylacrylic acid, 2-fluoro-3-phenyl-2-propenoic acid, 2z-2-fluoro-3-phenylprop-2-enoic acid, 2-fluoro-3-phenylacrylic acid, cinnamic acid, .alpha.-fluoro
PubChem CID 1550911
IUPAC Name (Z)-2-fluoro-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=C(C(=O)O)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 156°C to 160°C
Color White
Infrared Spectrum Authentic
Linear Formula C6H5CH=C(F)CO2H
Quantity 1 g
Beilstein 09, I, 237
Formula Weight 166.15
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material α-Fluorocinnamic acid, 98%
Show More Show Less
EINECSNumber 206-508-0
RTECSNumber GD9080000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.