Learn More
Crescent Chemical Co Inc FLUOROMOUNT W FOR MICROSCOPY
SDP
Supplier:  Crescent Chemical Co Inc SE21634.01
Encompass_Preferred
Fluoromount W for microscopy aqueous solution Non-fluorescent mounting medium for microscopy. Ideal for F.I.T.C. Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 21634.01
Catalog No. 50-247-596