Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fluoroshield Mounting Medium, Aqueous, Liquid, 1000 mL

Catalog No. 502585556
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5556 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5556 Supplier Abcam Supplier No. AB1041351000ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aqueous mounting medium formulated to prevent rapid photobleaching and preserve fluorescence in tissue and cell imaging.

  • Aqueous mounting medium designed to preserve fluorescence for tissue and cell sample imaging
  • Prevents rapid photobleaching of key fluorophores
  • Fluorescence remains stable at 4°C in the dark
  • Cited in over 30 publications
  • Form: Liquid
  • Constituents: 84% Water
  • Preservative: 0.05% Sodium azide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Blue Ice
Buffer 84% water, 0.05% sodium azide
Form Liquid
Detection Method Fluorescence
Content And Storage Store at 4°C in the dark.
Formulation 84% water, 0.05% sodium azide
Sample Type Tissues, Cell samples
Quantity 1000 mL
For Use With (Application) Fluorescence imaging of tissue and cell samples
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.