Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Drop Plate for Fluoroskan™ FL Microplate Fluorometer and Luminometer

Catalog No. 50950290
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Fluoroskan
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-950-290 Fluoroskan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-950-290 Supplier Thermo Scientific™ Supplier No. 2805880
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Perform a number of fluorometric and luminometric research applications with the Thermo Scientific™ Fluoroskan Ascent™ FL Microplate Fluorometer and Luminometer.

Perform a number of fluorometric and luminometric research applications with the Thermo Scientific™ Fluoroskan Ascent™ FL Microplate Fluorometer and Luminometer, which offers versatile plate formats, fast plate reading speeds, top/bottom plate reading and up to three dispensers. Compact and robust, the excellent optical performance of this machine also allows it to perform ideally on life science research applications.

Fast reading speed:

  • Reads a 96-well plate in just 15 seconds, which is essential for kinetic applications.
  • Combination of fluorometric GFP measurement and a lumometric luciferase measurement from the same well is fast due to the versatility of assay programming.

Onboard dispensers for exact follow-up of kinetic reactions:

  • Equipped with up to three reagent dispensers for rapid kinetic assays, such as Ca2+ flux
  • Supports simultaneous dispensing and reading, enabling monitoring of kinetic measurements from the very start of the reaction
  • Very low dead volume and the backflush capability for conservation of expensive reagent

High sensitivity for both top and bottom reading:

  • Fiberless direct illumination optics for both top and bottom reading
  • High sensitivity, wide dynamic range and low cross talk for accurate and precise results for 1- to 384-well plates

Specially designed for automation:

  • Reads up to 384-well plates for increased assay throughput
  • Easily integrated with automated systems

Includes:

  • PC Software and filter pairs:
    • Excitation: 355 nm, Emission: 460 nm
    • Excitation: 485 nm, Emission: 538 nm
    • Other filters available upon request

Recommended for:

Ca2+ flux assays, Cell proliferation, Cytotoxicity, Cell adhesion, DNA quantitation, Reporter gene assays, Hybridization assays, Ion Channel, Quantitation of PCR products, FRET assays, BRET assays, Molecular beacon assays, Immunoassays, Enzyme activity, Bacterial quantitation, Phagocytosis, Oligonucleotide assays

TRUSTED_SUSTAINABILITY

Specifications

Product Type Drop Plate
For Use With (Equipment) Fluoroskan
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.