Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FluoSpheres™ Sulfate-Modified Microspheres, 0.02–4.0 μm

Catalog No. F8858
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
0.02 μm
0.20 μm
1 μm
2 μm
4 μm
Color:
Red
Yellow-Green
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Color
F8858 4 μm Red
F8851 1 μm Red
F8845 0.02 μm Yellow-Green
F8848 0.20 μm Yellow-Green
F8852 1 μm Yellow-Green
F8853 2 μm Yellow-Green
F8859 4 μm Yellow-Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. F8858 Supplier Invitrogen™ Supplier No. F8858
Add to Cart
Edge
Add to Cart

Perform flow cytometry, microscopy, HTS, and HCS with our FluoSpheres Sulfate-Modified Microspheres, featuring different bead diameters.

Easily perform flow cytometry, microscopy, HTS, HCS, and other laboratory applications using our extensive selection of FluoSpheres Sulfate-Modified Microspheres. These microsphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications.

FluoSpheres Sulfate-Modified Microspheres are loaded with our proprietary fluorescent dyes, making them the brightest microspheres available. Because the fluorescent dye molecules are contained inside each polystyrene microsphere instead of only on the surface, the dye molecules are shielded from many of the environmental effects that cause quenching or photobleaching of exposed fluorophores. The stability, uniformity, and reproducibility of FluoSpheres microspheres, as well as their extensive color selection, make them the preferred tools for research and diagnostic assays that use fluorescence.

FluoSpheres Sulfate-Modified Microspheres are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin.

Key applications of FluoSpheres Sulfate-Modified Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

FluoSpheres sulfate-modified microsphere specifications for F8845, F8848, F8852, F8853, and F8859:
• Label (excitation/emission): yellow-green (505/515)
• Nominal bead diameters: 0.02, 0.2, 1.0, 2.0, and 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres sulfate-modified microsphere specifications for F8851 and F8858:
• Red (580/605)
• Nominal bead diameters: 1.0, 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres fluorescent microspheres are potentially more sensitive than colorimetric methods in most, if not all, major microsphere-based diagnostic test systems presently in use, including microsphere-agglutination tests, filter separation tests, particle-capture ELISA methods, and two-particle sandwich techniques.

FluoSphere Sulfate-Modified Microspheres (also called latex beads or latex particles) are manufactured using high-quality, ultra-clean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. Every possible precaution is taken throughout the manufacturing process to ensure that the microparticles are kept free of contaminating agents. The final product is sold as a suspension in ultrapure water, in most cases containing 2 mM azide or 0.02% thimerosal as a preservative.

Other surface modifications are available to facilitate the coupling of various molecules and proteins to the surface of the bead, including amine- and carboxylate-modified FluoSphere microspheres.

Choices for unstained microspheres
We also offer hundreds of choices for ultra-clean surfactant-free microspheres for research and commercial applications.

Custom microspheres
We prepare custom microspheres upon request. For example, FluoSpheres Sulfate-Modified microsphere beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Color Red
For Use With (Application) Cell Analysis
Material Polystyrene
Product Type Microsphere
Product Line FLUOSPHERES
Quantity 2 mL
Shipping Condition Room Temperature
Surface Modification Sulfate
Volume (Metric) 2 mL
Diameter (Metric) 4 μm
Show More Show Less
I have some FluoSpheres polystyrene microspheres, with 20 nm diameter. They are aggregating a lot. What can I do about it?

The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).

I sonicated my 2.0 µm carboxylate-modified microspheres, as recommended, but saw foaming (bubbles) on top of the solution. Should I be concerned?

Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

I accidentally froze my microspheres; can I still use them?

Even brief freezing can cause irreversible aggregation and potential distortion of the bead shape. You should not use these microspheres.

My protein-coated microspheres appear to be non-specifically binding in my experimental system. Do you have a product that can help reduce these non-specific interactions?

Non-specific binding can often be relieved by a blocking solution, but microspheres seem to require a stronger blocking solution than those most commonly commercially available. Hence, we've developed the BlockAid Blocking Solution (Cat. No. B10710). This reagent is a protein-based blocking solution designed for use with FluoSpheres microspheres and TransFluoSpheres microspheres conjugated to biotin, streptavidin, NeutrAvidin biotin-binding protein, or other proteins. The BlockAid Blocking Solution has proven useful for reducing the nonspecific binding of protein-coated or other macromolecule-coated microspheres in a wide variety of flow cytometry, microscopy, and microarray applications.

What sort of sonication method should I use to disperse microsphere aggregates?

We recommend using a bath sonicator to disperse microsphere aggregates. Do not use a probe sonicator as this will damage the microspheres.

I sonicated my carboxylate-modified latex microspheres as recommended in the protocol, and I saw foaming in the solution. Should I be concerned?

The foaming is from Tween 20, which is present in the stock solution to help prevent aggregation. It is normal to see bubbles from this reagent.

Can I centrifuge my fluorescent microspheres?

Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm, but once pelleted they may not disperse readily or not at all. Centrifugation is not an effective way to collect smaller microspheres; for beads less than 1 µm in diameter, we recommend washing by either cross-flow filtration or dialysis with a 500 kDa MWCO.

Which microspheres should I use to measure regional blood flow? How do I perform these procedures?

We offer a variety of FluoSphere microspheres and kits specifically designed for blood flow determination; if the available colors and/or sizes do not suit your needs, we recommend using beads modified with carboxylate groups for blood flow determination applications. We give some basic usage guidelines in the FluoSpheres Fluorescent Micospheres for Tracer Studies product manual (https://tools.thermofisher.com/content/sfs/manuals/mp13080.pdf). Additionally, the University of Washington Seattle offers the Fluorescent Microsphere Resource Center (FMRC) website (http://fmrc.pulmcc.washington.edu/fmrc.shtml) with detailed applications manuals and references for using fluorescent microspheres to evaluate regional blood flow.

What microsphere product should I select to perform a point spread function?

Point spread functions will require a subresolution bead, and we have two different subresolution bead products that could be used for this purpose. TetraSpeck Microspheres, 0.1 µm (Cat. No. T7279), are single subresolution microspheres that are simultaneously stained with blue, green, orange, and dark red dyes. The PS-Speck Microscope Point Source Kit (Cat. No. P7220) consists of individually-stained blue, green, orange, and dark red fluorescent microspheres that are approximately 175 nm in diameter.

What sort of mounting media should I use for fluorescent polystyrene microspheres?

We recommend using a ProLong product or any other aqueous mounting media. You should avoid organic-based mountants such as toluene-based mounting media.

Are the FluoSpheres microspheres surfactant-free?

The FluoSpheres microspheres may have a trace amount of surfactant. Only IDC Latex beads with diameters greater than 100 nm are actually surfactant-free.

I want to put my microspheres in something besides water; how will they react in alcohol?

The stability in alcohol or water/alcohol mixtures is dependent on the alcohol chain length. The beads are fairly stable in 40% methanol, but they will dissolve much more readily in long chain alcohols. Prolonged storage of beads in an alcohol containing solution will tend to leach out the dye.

I noticed that many of your microsphere products have sodium azide present, but I can't have sodium azide in my application. How should I remove sodium azide from the microsphere solution?

We recommend using dialysis with a MWCO of approximately 500,000 to remove the sodium azide from the microsphere-containing buffer.

How many dye molecules are there in a single fluorescent microsphere?

We do not determine the exact number of dye molecules per microsphere. The number of dye molecules is a function of the diameter of the particle and the properties of the dye.

How do you label the FluoSpheres microspheres with dye?

The polystyrene microspheres are swelled in a solvent containing a hydrophobic dye specific to the FluoSpheres product. The dye is then able to diffuse into the polystyrene matrix. The beads are then removed from the solvent and dialyzed into an aqueous environment; this reverses the swelling and traps the dye in the polystyrene. Once trapped, the dye is fairly protected from the external environment. The exact protocol and identity of the hydrophobic dyes are proprietary.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.