Learn More
Abcam Fluoxetine Hydrochloride, 5-HT Reuptake Inhibitor, >99% Purity, 10 mg
Description
- Molecular weight: 345.8 Da
- Purity: >99%
- Form: Solid
- Activity: Potent, selective 5-HT reuptake inhibitor. Antidepressant in vivo.
- Binding affinity: Binds to rat and human 5-HT transporters with Ki values of 2 and 0.9 to 20 nM, respectively.
- Selectivity: Shows approximately 150 to 900-fold selectivity over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic and muscarinic receptors.
- Storage: Store at +4°C.
- Citation count: 6
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 5-HT Transporter |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Reactivity | Rat, Human |
| Shipping Condition | Ambient |
| Quantity | 10 mg |
| For Use With (Application) | Functional studies, antidepressant research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.