Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fluoxetine Hydrochloride, 5-HT Reuptake Inhibitor, Solid, >99% Purity, 50 mg

Catalog No. 502707881
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7881 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7881 Supplier Abcam Supplier No. AB12007750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and selective 5-HT reuptake inhibitor with >99% purity.

  • Potency: Potent, selective 5-HT reuptake inhibitor.
  • Molecular Weight: 345.8 Da.
  • Purity: >99%.
  • Biological Activity: Antidepressant in vivo; binds to rat and human 5-HT transporters with Ki values of 2 and 0.9–20 nM, respectively.
  • Selectivity: Approximately 150–900-fold selectivity over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic and muscarinic receptors.
  • Form: Solid.
  • Storage: Store at +4°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule 5-HT Transporter
Form Solid
Buffer 100% Fluoxetine hydrochloride
Content And Storage Store at 4°C.
Cross Reactivity Rat, Human
Formulation 100% Fluoxetine hydrochloride
Quantity 50 mg
For Use With (Application) Functional studies, antidepressant research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.