Learn More
Apexbio Technology LLC Pramipexole 2HCl Monohydrate 191217-81-9 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B14875.1
Pramipexole 2HCl Monohydrate is a synthetic non-ergoline dopamine receptor agonist targeting dopamine receptor subtypes particularly D3 D2S D2L and D4 It is designed to activate these receptors thereby modulating dopaminergic transmission pathways Pramipexole 2HCl Monohydrate exerts its biological activity primarily through selective receptor activation Based on these pharmacological properties Pramipexole 2HCl Monohydrate holds research potential in studies of receptor-ligand interactions receptor subtype functional analysis and dopamine-related neurologic or psychiatric disorders including Parkinson s disease and restless legs syndrome
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.