Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fluticasone propionate
GREENER_CHOICE

Catalog No. 200710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-071-0 10 mg
20-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-071-0 Supplier Tocris Bioscience™ Supplier No. 2007/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective high affinity glucocorticoid receptor agonist

High affinity, selective glucocorticoid receptor agonist (Kd = 0.5 nM). Potently stimulates glucocorticoid receptor-mediated transactivation of gene expression and enhances human eosinophil apoptosis (EC50 = 3.7 nM) in vitro. Inhibits mast cell accumulation in nasal mucosa following topical administration. Lipophilic anti-inflammatory agent with low oral bioavailability. Also potentiates KV1 channels (EC50 = 37 nM).

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S

Specifications

Quantity 10 mg
Formula Weight 500.57
Purity >98%
Molecular Weight (g/mol) 505.604
Chemical Name or Material Fluticasone propionate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.