Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLVCR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528463 Supplier Invitrogen™ Supplier No. PA528463
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: FLVCR tansfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the major facilitator superfamily of transporter proteins. The encoded protein is a heme transporter that may play a critical role in erythropoiesis by protecting developing erythroid cells from heme toxicity. This gene may play a role in posterior column ataxia with retinitis pigmentosa and the hematological disorder Diamond-Blackfan syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLVCR1
Applications Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FLVCR1
Gene Accession No. Q9Y5Y0
Gene Alias 9630055N22Rik; AXPC1; eline leukemia virus subgroup C cellular receptor 1; feline leukemia virus subgroup C cellular receptor 1; feline leukemia virus subgroup C receptor; Feline leukemia virus subgroup C receptor-related protein 1; FLVCR; FLVCR heme transporter 1; FLVCR protein; FLVCR1; Major facilitator superfamily domain containing 7B; Mfsd7b; PCA; PCARP
Gene Symbols FLVCR1
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human FLVCR. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.