Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595285
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595285 Supplier Invitrogen™ Supplier No. PA595285
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human hepatocellular carcinoma tumor tissue, human HepG2 whole cell, human HUH7 whole cell, rat liver tissue, mouse liver tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMO1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and no preservative
Gene FMO1
Gene Accession No. P36365, P50285, Q01740
Gene Alias dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; fetal hepatic flavin-containing monooxygenase 1; flavin containing dimethylaniline monoxygenase 1; flavin containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; FMO 1A1; FMO form 1; Fmo1; Fmo-1; hepatic flavin-containing monooxygenase (EC 1.14.13.8); hepatic flavin-containing monooxygenase (FMO); hepatic flavin-containing monooxygenase 1; monooxygenase (FMO) (EC 1.14.13.8); RFMO1A
Gene Symbols FMO1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human FMO1 (334-363aa AFPFLDESVVKVEDGQASLYKYIFPAHLQK).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14261, 2326, 25256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.