Learn More
Sigma Organic Chemistry Fmoc-1-aminomethyl-cycl | 10mg | MFCD09264208
Supplier: Sigma Organic Chemistry CDS00712810MG

Fmoc-1-aminomethyl-cycl, 10mg
About this Item:
MDL #: MFCD09264208
Molecular Weight: 379.45
UNSPSC Code: 12352200
Chemical Formula: C23H25NO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.