Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-2-Nal-OH, Advanced ChemTech

Catalog No. 01340763
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
01-340-763 25 g
01-340-762 5 g
01-340-764 1 g
3 options

Catalog No. 01-340-763

Supplier: Advanced Chem Tech CB720825G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112883-43-9
Molecular Formula C28H22NO4
Molecular Weight (g/mol) 436.49
InChI Key JYUTZJVERLGMQZ-SANMLTNESA-M
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoate
SMILES [O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-2-Nal-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.