Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Arg(Pbf)-OH 98.0+%, TCI America™
SDP

Catalog No. F072925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F072925G 25 g
F07295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F072925G Supplier TCI America Supplier No. F072925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 154445-77-9
Molecular Formula C34H40N4O7S
Molecular Weight (g/mol) 648.775
MDL Number MFCD00235804
InChI Key HNICLNKVURBTKV-NDEPHWFRSA-N
Synonym fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4
PubChem CID 11354259
IUPAC Name (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 648.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.