Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Cys Trt -OPfp, Advanced ChemTech

Catalog No. 01340600
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-600 100 g
01-340-597 5 g
01-340-598 25 g
01-340-599 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-340-600 Supplier Advanced Chem Tech Supplier No. FC5214100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115520-21-3
Molecular Formula C43H30F5NO4S
Molecular Weight (g/mol) 751.77
InChI Key FDAUCYDVXIPBDR-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoate
SMILES FC1=C(F)C(F)=C(OC(=O)C(CSC(C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(F)=C1F

Specifications

Color White/Off-White
Quantity 100 g
Grade Biochemical
Chemical Name or Material Fmoc-Cys Trt -OPfp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.