Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-D-Phe 4-Cl -OH, Advanced ChemTech

Catalog No. 01340431
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-340-431

Supplier: Advanced Chem Tech FF34291G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142994-19-2
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.88
InChI Key CQPNKLNINBUUOM-UHFFFAOYNA-N
IUPAC Name 3-(4-chlorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES OC(=O)C(CC1=CC=C(Cl)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-D-Phe 4-Cl -OH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.