Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-D-Tyr(4-Et)-OH | 162502-65-0, 10GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1068310GM
Encompass
WuXi TIDES Fmoc-D-Tyr(4-Et)-OH, 162502-65-0 | (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid | CCOC1=CC=C(C[C@@H](NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)C(O)=O)C=C1 | M.F. C26H25NO5, M.W. 431.49
Catalog No. 50-236-0232