Learn More
Aat Bioquest FMOC-Lys(5-TAMRA)-OH
SDP
Supplier:  Aat Bioquest 5045
Encompass_Preferred
FMOC-Lys(5-TAMRA)-OH is a building block for in-sequence Lys labeling by TAMRA. TAMRA is one of the most common fluorescent dyes for preparing FRET peptides (often paired with FAM or TQ3).
Catalog No. 50-195-3262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.