Learn More
Bachem Fmoc-Thr(Trt)-OH
Supplier: Bachem 4025725.0005
Fmoc-Thr(Trt)-OH, 133180-01-5, C38H33NO5, Mr 583.68, 5g. The trityl group can be removed by 1% TFA, leaving other protecting groups intact. On-resin Trt cleavage was employed by Biron et al. in a synthesis of 1,3-oxazole-containing peptides.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.