Learn More
MilliporeSigma FMOC-TYR(TBU)-OH
Supplier:  MilliporeSigma 8520200100
Encompass
Fmoc-Tyr(tBu)-OH; > /=98% by HPLC and TLC; White to light yellowish solid powder; EMD; Novabiochem; 100g; N-alpha-Fmoc-O-tert-butyl-L-tyrosine; F.W. 459.6; C28H29NO5; M.P. 143-155 deg.C; CAS: 7
Catalog No. 50-230-1680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.