Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMR1 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
SDP

Catalog No. 501675013
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5013 100 μL
50-254-0407 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5013 Supplier OriGene Supplier No. TA504293
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FMR1 binds RNA and is associated with polysomes. The protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat in the 5′ UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure. Multiple alternatively spliced transcript variants that encode different protein isoforms and which are located in different cellular locations have been described for this gene.

Specifications

Antigen FMR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C3
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FMR1
Gene Accession No. P35922, Q06787, Q80WE1
Gene Alias FMRP, FRAXA, POF, POF1
Gene Symbols FMR1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 26-279 of human FMR1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14265, 2332, 24948, 481070
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.