Learn More
CPC Scientific H-Phe-Met-Arg-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific FMRF002A
Encompass
Molluscan neuropeptide cardioexcitatory and lyophilized solid that localizes with neuropeptide Y in some regions of the brain. This peptide also inhibits Na+-Ca2+ exchange in cardiac sarcolemmal vesicles.SEQUENCE: H-Phe-Met-Arg-Phe-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: FMRF-NH2MOLECULAR FORMULA: C29H42N8O4S1MOLECULAR WEIGHT: 598.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [152165-14-5]SYNONYMS: FMRF-amide, Molluscan Cardioexcitatory Neuropeptide, FMRFamide, H-Phe-Met-Arg-Phe-NHRESEARCH AREA: NeuropeptidesREFERENCES:D.A. Price et al., Science, 197, 670 (1977)SKU(s): FMRF-002A, FMRF-002B
Catalog No. 50-210-9302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.