Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMRP Recombinant Superclonal™ Antibody (5HCLC)

Catalog No. PI711441 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711441 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711441 Supplier Invitrogen™ Supplier No. 711441
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Cat, Pig and Sheep Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

FMR1 binds RNA and is associated with polysomes. The protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat in the 5' UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure. Multiple alternatively spliced transcript variants that encode different protein isoforms and which are located in different cellular locations have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMRP
Applications Immunocytochemistry
Classification Recombinant Superclonal
Clone 5HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene FMR1
Gene Accession No. Q06787
Gene Alias AT24755; BcDNA:GM08679; cg 6203 gene product from transcript cg6203-rc; cg6203; CG6203-PA; CG6203-PB; CG6203-PC; CG6203-PD; CG6203-PE; CG6203-PF; CG6203-PG; CG6203-PH; CG6203-PI; CG6203-PJ; CG6203-PK; dFMR; DFmr1; dFmrp; dfxr; dfxr1; dFXRP; Dmel\CG6203; Dmel_CG6203; dmfr1; drosophila fragile X mental retardation protein; EP(3)3517; FMR; FMR1; Fmr-1; Fmr1-PA; Fmr1-PB; Fmr1-PC; Fmr1-PD; Fmr1-PE; Fmr1-PF; Fmr1-PG; Fmr1-PH; Fmr1-PI; Fmr1-PJ; Fmr1-PK; FMRP; FMRP translational regulator 1; fragile X; fragile X mental retardation; fragile X mental retardation 1; fragile X mental retardation gene; fragile X mental retardation protein; fragile X mental retardation protein 1; Fragile X mental retardation protein 1 homolog; fragile X mental retardation related 1; fragile X mental retardation syndrome 1; fragile X mental retardation syndrome 1 homolog; Fragile X mental retardation syndrome-related protein 1; fragile X mental retardation-1 protein; fragile X protein; fragile x related; fragile X related protein; fragile X retardation 1 protein; fragile X-related; fragile-X; Fragile-X mental retardation 1; Fragile-X mental retardation protein; Fragile-X-related; FRAXA; FXR; MGC87458; POF; POF1; protein FMR-1; ragile X mental retardation protein; synaptic functional regulator FMR1
Gene Symbols FMR1
Host Species Rabbit
Immunogen Protein corresponding to Human FMR1 (216-425).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.