Learn More
Enzo Life Sciences Fibronectin (plasma) (human) monoclonal antibody (BF12) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLFG60200100
Encompass_Preferred
Clone: BF12. Isotype: IgG1. Immunogen: Purified fibronectin from A8387 fibrosarcoma cells. Species reactivity: Human. Specificity: Recognizes the cell binding region of fibronectin. Applications: ICC, WB. Purity: Protein A affinity chromatography. Formulation: Liquid. In PBS containing 0.1% sodium azide and 1mg/mL BSA.
Catalog No. 50-201-0823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.