Learn More
AdipoGen Fn14 (human):Fc (human) (rec.) (3 x 50 µg)
SDP
Supplier:  AdipoGen AG40B00343050
Encompass_Preferred
Protein. The extracellular region of human FN14 (aa 28-75) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to mouse and human TWEAK. Purity: >95% (SDS-PAGE). TWEAK is a cytokine that controls proliferation, migration, differentiation, apoptosis, angiogenesis and inflammation. TWEAK binds to Fn14, a highly inducible cell-surface receptor that is linked to several intracellular signalling pathways, including the nuclear factor-B (NF-kappaB) pathway. The TWEAK–Fn14 axis plays a beneficial role in tissue repair following acute injury and may contribute to cancer, chronic autoimmune diseases and acute ischaemic stroke.
Catalog No. 50-205-3105
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.