Learn More
Newcomer Supply FORM-Zero Formalin Neutralizer, 500cc bottle, 12/case, Powder Reagent
Supplier: Newcomer Supply ABFZ55012
Newcomer Supply Form–Zero™ Formalin Neutralizer is a unique powdered reagent blend of various sulfur-containing inorganic salts. Form–Zero™ readily dissolves in aqueous solutions and subsequently functions as a chemical reducing agent to quickly neutralize formalin and glutaraldehyde for sink disposal. One 500 cc bottle of Form–Zero™ will neutralize one gallon/four liters of 10% formalin or one gallon/four liters of 4% glutaraldehyde. Benefits include: No pH adjustment of solutions is required, One gallon/four liters of formalin or glutaraldehyde can be neutralized in 20-25 minutes to non-hazardous disposable solutions, Neutralized aldehyde products can be safely discharged to sewer and waste water treatment systems and No drain clogging polymers are created.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.