Learn More
Bellco Glass, Inc. Foam Level Probes are used in fermentation bioreactors to detect the presence of foam or the liquid level within the bioreactor vessel. When the foam level or liquid level reaches the tip of the probe, a conductance path is created for the probe to trigge
SDP
Supplier:  Bellco Glass, Inc. 780341600
Encompass_Preferred
Foam Level Probes are used in fermentation bioreactors to detect the presence of foam or the liquid level within the bioreactor vessel. When the foam level or liquid level reaches the tip of the probe, a conductance path is created for the probe to trigge
Catalog No. 50-194-6337
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.