Learn More
Grainger Horn Finish Red Flush
Supplier:  Grainger 16X364
Encompass_Preferred
The System Sensor 14N226 is a horn that produces a loud, piercing sound to alert people when a fire or security alarm is triggered. This 12 volt DC electronic horn has a sound output of 88 decibels at 10 feet and is rated for indoor use. The horn is made of durable red plastic with dimensions of 5.2 x 3 x 2.5 inches. It can be mounted to walls or ceilings using the included mounting bracket and screws. The 14N226 horn is ETL listed and meets the requirements of NFPA 72 for audible signaling appliances.
Catalog No. 50-276-6182
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.