Learn More
Foamtec PharmaMOP™ Cleanroom Mops, Sterile
Catalog No. 19304832 Shop All Foamtec International Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
19-304-832 Sterile
1 options
Catalog No. 19-304-832 Supplier Foamtec Supplier No. FS4500PHMST
For cleaning and disinfection of all types of critical surfaces
  • Gamma irradiated
  • Polyester/microfiber made mop head
  • Locking head frame, 5.25 ft. telescoping handle
Specifications
Sterility Sterile