Description
- Removes deposition from areas critical to process
- Prevents unwanted flaking
- High impact plastic w/spring-tensioned attachment mechanism
- Interchangeable belt-shaped
- Spring adjustable for easy mounting and rotation
- Minimizes tool wear
- For polishing stains, scratches and process-induced residue from scratch sensitive surfaces
Specifications
Specifications
| Product Type | Scrub Belt |
| For Use With (Application) | For quick removal of deposition from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces |
| Grit | 280 |
| Material | Diamond |