Learn More
Foamtec Stainless Steel Corrosion Remediation Kits
Catalog No. 19304556 Shop All Foamtec International Products
Change view
Click to view available options
Product Type:
Large Surface Starter Kit
Large Surface Starter Scrubpad Refill Kit
Mini Kit
Natural Sahara Sponge
Small Surface Starter Kit
Small Surface Starter Scrubpad Refill Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-304-556 Small Surface Starter Kit
19-304-555 Large Surface Starter Kit
19-304-812 Small Surface Starter Scrubpad Refill Kit
19-304-813 Large Surface Starter Scrubpad Refill Kit
19-304-581 Mini Kit
19-305-775 Natural Sahara Sponge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-304-556 Supplier Foamtec Supplier No. HT4500SSC2
This item is not returnable. View return policy
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
Kits include polishing and cleaning products along with instructional videos and documentation that result in mirror smooth 304 and 316 SS surfaces that resist subsequent corrosion.
Kit Contents
19-304-555: Stainless Steel Corrosion Remediation Starter Kit for Large Surfaces
2 Handles, 10 Diamond ScrubDISKS (5 girts), 1 Silicon-Carbide ScrubDISK, 1PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 25 MiraWIPE Wipers, 2 UltraSOLV Sponge, and Flash Drive with instructional Videos
19-304-556: Stainless Steel Corrosion Remediation Starter Kit
1 PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 10 MiraWIPE Wipers, 1 UltraSOLV Sponge, and Flash Drive with instructional Videos
Specifications
Product Type Small Surface Starter Kit
For Use With (Application) Cleaning corrosion on stainless steel surfaces
For Use With (Equipment) Sahara Corrosion Cleaning System
Includes 280 Diamond grit ScrubPAD, 360 Diamond grit ScrubPAD, 1350 Diamond grit ScrubPAD, 1500 Silicon Carbide grit ScrubPAD, 2000 Diamond grit ScrubPAD, 280 Diamond grit ScrubDISK™, 360 Diamond grit ScrubDISK™, FTP