Description
- Tear-resistant, highly absorbent polyurethane sponge
- With stainless steel bracket
- Pin-hole-free foam for abrasion resistance
- Available in Bump-Proof™ micro fiber over foam versions
- For wet mopping procedures in sterile and controlled environments
Specifications
Specifications
| Product Type | Self-wringing Sponge Mop Head |
| Length (English) | 14 in. |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Quantity | 12/Cs. |
| Includes | Wrapped with Polyester Lamination, Stainless Steel Bracket |
| For Use With (Application) | Designed for wet mopping procedures in sterile and controlled environments |
| Material | Polyurethane |