Learn More
Andwin Scientific FOIL SEALING SQUARES 8X8
SDP
Supplier:  Andwin Scientific EF9895C
Encompass_Preferred
The Aluminum Foil Sealing Squares can be utilized for autoclaving and incubating. This product can resist tears and punctures and can mold to fit and completely cover openings. The squares fit 8 x 8 inches.
Catalog No. NC9630961
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.