Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folic Acid, Fisher BioReagents

Catalog No. BP251910 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP251910 10 g Amber Glass
BP25195 5 g Amber Glass
BP251925 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP251910
Folic Acid is used in cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Melting Point 250°C
Color Yellow to Orange
Density 1.59g/cm³
pH 4.0 to 4.8
Ignition Residue 0.3% max.
Loss on Drying 8.5% max.
Assay Percent Range ≥95.0 %
Packaging Amber Glass
Quantity 10 g
Merck Index 15, 4248
Formula Weight 441.39
Percent Purity 95.0% min.
Physical Form Powder or Solid
Chemical Name or Material Folic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT, protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.