Learn More
Description
Folin and Ciocalteu's phenol reagent has been used for the estimation of total phenolic content in samples.
Specifications
Specifications
| Chemical Name or Material | Folin and Ciocalteu’s Phenol Reagent |
| Concentration | 1.9 to 2.1 N |
| Packaging | Glass Bottle |
| pH | <0.5 |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.