Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque FOLR1 Protein, His tag
Supplier:  ACROBiosystems FO1C52H81MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 35-41 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7045
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.