Learn More
Genscript Corporation MSLN/Mesothelin306-590 hFc C
Supplier:  Genscript Corporation Z03874500
Encompass_Preferred
Mesothelin also known as MSLN is a protein that in humans is encoded by the MSLN gene Cloning studies showed that the mesothelin gene encodes a precursor protein that is processed to yield mesothelin which is attached to the cell membrane by a glycophosphatidylinositol linkage and a 31-kDa shed fragment named megakaryocyte-potentiating factor (MPF) Although it has been proposed that mesothelin may be involved in cell adhesion its biological function is not known A knockout mouse line that lacks mesothelin reproduces and develops normally
Catalog No. 50-000-56720
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.