Learn More
ACROBiosystems Recombinant Protein;Human FOLR1 Protein, Strep Tag;HEK293;250UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Supplier:  ACROBiosystems FO1H528B250UG
Encompass_Preferred
Recombinant Protein;Human FOLR1 Protein, Strep Tag;250UG;Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_057937.1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8623
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.