Learn More
Description
FOLR2 Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | FOLR2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4G6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FOLR2 |
| Gene Accession No. | P14207 |
| Gene Alias | BETA-HFR; FBP; FBP/PL-1; Fbp2; folate binding protein; folate binding protein 2; folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
