Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR3 Mouse anti-Human, Clone: OTI7E10, liquid, TrueMAB™
SDP

Catalog No. 501684107
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4107 Supplier OriGene Supplier No. TA811544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Folate is an essential vitamin that must be obtained from food intake through intestinal absorption in mammals. Folate and reduced folic acid derivatives bind to the folate receptor (FR) family, which mediates the endocytosis of 5-methyltetrahydrofolate into the cell. The folate receptors consist of five members, alpha, beta, gamma, gamma′ (which is produced by alternative splicing) and delta. beta-FR is attached to the membrane by a GPI anchor and is expressed in maligt tissues of epithelial and nonepithelial origin. gamma-FR is expressed in tissues of hematopoietic origin, such as spleen, thymus and bone marrow. The expression pattern of delta-FR is elusive, which suggests that it is highly restricted both spatially and temporally. The alpha isoform of the folate receptor is used as a highly selective tumor marker and may be targeted for the delivery of therapeutic compounds to tumor cells by coupling to derivatives of folic acid.

Specifications

Antigen FOLR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7E10
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FOLR3
Gene Accession No. P41439
Gene Alias FR-G, FR-gamma, gamma-hFR
Gene Symbols FOLR3
Host Species Mouse
Immunogen Full length human recombit protein of human FOLR3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2352
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.